Learn More
Biomatik Corporation Mouse Breast Cancer Susceptibility Protein 2 (BRCA2) ELISA Kit
Supplier:  Biomatik Corporation EKU02809
Encompass_Preferred
Mouse Breast Cancer Susceptibility Protein 2 (BRCA2) ELISA Kit for detecting BRCA2 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97929. Gene ID: 12190. Target Synonyms: BRCA2; BRCC2; FACD; FAD; FAD1; FANCD; FANCD1; Breast Cancer 2,Early Onset; Breast Cancer Type 2 Susceptibility Protein; Fanconi anemia group D1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5522