Learn More
Sino Biological Mouse BSG MAb 20ul (Sino Biological)
Supplier:  Sino Biological 50332R04120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD147 (rM CD147; Catalog#50332-M08H; NP_001070652.1; Met 1-Arg 209).
Catalog No. 50-237-3405