Learn More
Sino Biological Anti-CD147/Basigin/EMMPRIN Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 109373T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse CD147, and purified by antigen affinity chromatography.
Catalog No. 50-255-5046