Learn More
Raybiotech Inc Mouse Tryptase epsilon ELISA Kit
Supplier:  Raybiotech Inc ELMBSSP41
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 60 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects mouse BSSP-4. Other species not determined.
Catalog No. 50-194-9937