Learn More
Sino Biological Recombinant Mouse BTLA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 51060M08H1
Encompass_Preferred
A DNA sequence encoding the Mouse BTLA (AAI08964.1) (Met1-Gly176) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-240-9387