Learn More
American Research Products Inc Mouse C C chemokine receptor t
Catalog No. 5000327534
Encompass
Mouse C-C chemokine receptor type 2 (CCR2) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27534 Supplier American Research Products Inc Supplier No. CSBEL004841MO
Mouse C-C chemokine receptor type 2 (CCR2) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich