Learn More
American Research Products Inc Mouse C C motif chemokine 25
Catalog No. 5000327133
Encompass
Mouse C-C motif chemokine 25 (CCL25) ELISA kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27133 Supplier American Research Products Inc Supplier No. CSBEL004789MO
Mouse C-C motif chemokine 25 (CCL25) ELISA kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich