Learn More
Description
PEP-242 is a 14 amino acid synthetic peptide corresponding to amino acid residues 1-14 from mouse C/EBP alpha. The sequence of this peptide is (amino to carboxy terminus): M E S A D F Y E V E P R P P. This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # MA1-825. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (~70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 mL of distilled water.
Specifications
Specifications
| Gene ID (Entrez) | 12606 |
| Species | Mouse |
| Purification Method | purified |
| Accession Number | P53566 |
| Common Name | C/EBP alpha |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | Peptide with no preservative |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.