Learn More
American Research Products Inc Mouse C ReactiVe Protein CRP
Catalog No. 5000326095
Encompass
Mouse C-ReactiVe Protein CRP ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26095 Supplier American Research Products Inc Supplier No. CSBE07923M
Mouse C-ReactiVe Protein CRP ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich