Learn More
American Research Products Inc Mouse C ReactiVe Protein CRP
Supplier:  American Research Products Inc CSBE07923M1
Encompass
Mouse C-ReactiVe Protein CRP ELISA Kit from CUSABIO Detection Range 5 ng/mL-4000 ng/mL Sensitivity 5 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26254