Learn More
Sino Biological Mouse C-Reactive Protein Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785092
Encompass_Preferred
The Mouse C-Reactive Protein/CRP ELISA Pair Set is for the quantitative determination of Mouse C-Reactive Protein/CRP.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse C-Reactive Protein/CRP coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse C-Reactive Protein/CRP present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse C-Reactive Protein/CRP polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse C-Reactive Protein/CRP present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85092 Supplier Sino Biological Supplier No. SEK5040915
The Mouse C-Reactive Protein/CRP ELISA Pair Set is for the quantitative determination of Mouse C-Reactive Protein/CRP.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse C-Reactive Protein/CRP coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse C-Reactive Protein/CRP present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse C-Reactive Protein/CRP polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse C-Reactive Protein/CRP present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.