Learn More
American Research Products Inc Mouse C X C motif chemokine 11
Catalog No. 5000327221
Encompass
Mouse C-X-C motif chemokine 11 (CXCL11) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27221 Supplier American Research Products Inc Supplier No. CSBEL006241MO
Mouse C-X-C motif chemokine 11 (CXCL11) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich