Learn More
American Research Products Inc Mouse C X C motif chemokine 13
Catalog No. 5000328578
Encompass
Mouse C-X-C motif chemokine 13 (CXcl13/Blc/Scyb13) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 096 ng/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich
Catalog No. 50-003-28578 Supplier American Research Products Inc Supplier No. CSBE16832M
Mouse C-X-C motif chemokine 13 (CXcl13/Blc/Scyb13) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 096 ng/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich