Learn More
American Research Products Inc Mouse C X C motif chemokine 14
Catalog No. 5000328234
Encompass
Mouse C-X-C motif chemokine 14 (CXCL14) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28234 Supplier American Research Products Inc Supplier No. CSBEL006244MO
Mouse C-X-C motif chemokine 14 (CXCL14) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma Method Sandwich