Learn More
American Research Products Inc Mouse C X C motif chemokine 3
Catalog No. 5000326876
Encompass
Mouse C-X-C motif chemokine 3 (CXCL3) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26876 Supplier American Research Products Inc Supplier No. CSBEL006249MO
Mouse C-X-C motif chemokine 3 (CXCL3) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich