Learn More
Complement Technology Inc M099 Mouse C1q Protein, Frozen liquid, 100 μg/vial, 1.0 mg/mL, > 85% purity, 10 mM HEPES, 300 mM NaCl, pH 7.2, -70°C storage, Fc domains of IgG or IgM binding
Supplier:  Complement Technology Inc M099
Encompass
M099 Mouse C1q Protein, Frozen liquid, 100 μg/vial, 1.0 mg/mL, > 85% purity, 10 mM HEPES, 300 mM NaCl, pH 7.2, -70°C storage, Fc domains of IgG or IgM binding
Catalog No. NC1652473