Learn More
BIOHIPPO Mouse C1ra Microsample
SDP
Catalog No. 5000315744
Encompass
Mouse C1ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of C1ra in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse C1ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse C1ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse C1ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15744 Supplier BIOHIPPO Supplier No. BHE15210615
Mouse C1ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of C1ra in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse C1ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse C1ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse C1ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color