Learn More
Abcam Mouse C3 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502656467
Encompass_Preferred
Mouse C3 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse C3.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6467 Supplier Abcam Supplier No. AB26770410X96TESTS
Mouse C3 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse C3.
The product is subject to the following: Abcam Restricted Use Statement