Learn More
Abcam Mouse C3 ELISA Kit, Sandwich ELISA, 96 tests, Colorimetric detection, Storage 4 °C, Sensitivity 25.52 pg/mL
Supplier:  Abcam AB2638841X96TESTS
Encompass_Preferred
The Abcam Mouse C3 SimpleStep ELISA Kit is designed for the quantitative measurement of mouse C3 protein in various biological samples. Utilizing a streamlined process, this kit provides results in just 90 minutes, ideal for complement system research. The technology allows for the formation of the antibody-analyte sandwich complex in a single, efficient step, enhancing usability and accuracy. This versatile kit is validated for multiple sample types including serum and plasma, making it suitable for a wide range of studies and applications.
  • Rapid quantification: Delivers results in 90 minutes with minimal steps.
  • High sensitivity: Detects down to 25.52 pg/mL.
  • Broad sample compatibility: Suitable for serum, plasma, and extracts.
  • Convenient format: Comes as a pre-coated 96-well microplate for ease of use.
Catalog No. NC1908872