Learn More
Sino Biological Recombinant Mouse Complement component 3 Protein (His Tag)
Supplier:  Sino Biological 56999M08H100
Encompass_Preferred
A DNA sequence encoding the Mouse C3 (NP_033908.2) (Ala994-Arg1310) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-241-0550