Learn More
Biomatik Corporation Mouse C3 (Complement C3) ELISA Kit
Supplier:  Biomatik Corporation EKF58702
Encompass_Preferred
Mouse C3 (Complement C3) ELISA Kit for detecting C3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01027. Target Synonyms: C3; ASP; CPAMD1; AHUS5; ARMD9; C3a; C3b. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6218