Learn More
Sino Biological C5a Protein, Mouse, Recombinant, Biotinylated
Supplier:  Sino Biological 51136MNAEB20
Encompass_Preferred
A DNA sequence encoding the mouse C5a (NP_034536.2) (Asn679-Arg755) was expressed with an initial Met. The purified protein was biotinylated in vitro.
Catalog No. 50-255-2982