Learn More
Lonza Walkersville Mouse C57 Brain Cortex Neurons
Supplier:  Lonza Walkersville MCX300
Encompass
Ready-to-use Clonetics Mouse Brain C57 Cortex Neurons (M-CX-300) are isolated from Mouse C57 black brain (E14, 15). These are sold as cell suspensions of high quality primary embryonic brain neuronal cells (including glia) prepared by standardized methods, and are ready for immediate culture.Benefits:Ready-To-Use No tissue dissection or animal handling required Increased data reproducibilityCharacteristic morphology and staining tests confirm tissue origin and cell typeNegative for mycoplasma, bacteria, yeast and fungiCertificate of Analysis (CoA) is available for every batchApplications:NeurogenesisNeurotransmitter FunctionGene ExpressionSignaling PathwaysElectrophysiologyNeurotoxicityDrug/Compound ScreeningAdvanced Cell Culture ModelsContent:≥4,000,000 cryopreserved cells per ampule
Catalog No. NC2034097