Learn More
Lonza Walkersville Mouse C57 Brain Mixed Astrocyt
Catalog No. 5000496392
Encompass
Ready-to-use Clonetics Mouse Brain Mixed Astrocytes (M-ASM-330) are obtained from C57 black mouse brain passaged once and prepared as cell suspensions for shipment Each vial contains approximately 1 million cells The astrocytes can be easily thawed and cultured Astrocytes can be harvested and re-plated The cells are shipped overnight on dry ice Recommended medium AGM BulletKit (CC-3186)
Catalog No. 50-004-96392 Supplier Lonza Walkersville Supplier No. MASM330
Add to Cart
Add to Cart
Ready-to-use Clonetics Mouse Brain Mixed Astrocytes (M-ASM-330) are obtained from C57 black mouse brain passaged once and prepared as cell suspensions for shipment Each vial contains approximately 1 million cells The astrocytes can be easily thawed and cultured Astrocytes can be harvested and re-plated The cells are shipped overnight on dry ice Recommended medium AGM BulletKit (CC-3186)