Learn More
Lonza Walkersville Mouse C57 Brain Striatum Neuro
Catalog No. 5000496370
Encompass
Ready-to-use Clonetics Mouse C57 Brain Striatum Neurons (M-CP-302) are isolated from mouse C57 black brain (E14 15) These are sold as cell suspensions of high quality primary embryonic brain neuronal cells (including glia) prepared by standardized methods and are ready for immediate culture Each vial of striatum cells contains approximately 4 million viable cells This will seed into 6 or more wells of a 24-well plate and 30 or more wells of a 96-well plate using the recommended plating densities and medium After approximately 4 days in culture the cells will form a neurite network and by the 7th day debris will be minimal Recommended medium PNGM BulletKit (CC-4461)
Catalog No. 50-004-96370 Supplier Lonza Walkersville Supplier No. MCP302
Ready-to-use Clonetics Mouse C57 Brain Striatum Neurons (M-CP-302) are isolated from mouse C57 black brain (E14 15) These are sold as cell suspensions of high quality primary embryonic brain neuronal cells (including glia) prepared by standardized methods and are ready for immediate culture Each vial of striatum cells contains approximately 4 million viable cells This will seed into 6 or more wells of a 24-well plate and 30 or more wells of a 96-well plate using the recommended plating densities and medium After approximately 4 days in culture the cells will form a neurite network and by the 7th day debris will be minimal Recommended medium PNGM BulletKit (CC-4461)