Learn More
BIOHIPPO Mouse C5a Microsample
SDP
Catalog No. 5000314041
Encompass
Mouse C5a Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of C5a in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 59 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse C5a Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse C5a Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse C5a biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14041 Supplier BIOHIPPO Supplier No. BHE15212331
Mouse C5a Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of C5a in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 59 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse C5a Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse C5a Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse C5a biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color