Learn More
ACROBiosystems Mouse CA125 / MUC16 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems CA5M52H6100UG
Encompass_Preferred
Mouse CA125 Protein, His Tag (CA5-M52H6) is expressed from human 293 cells (HEK293). It contains AA Asn 7750 - Pro 8422 (Accession # A0A140LJ72-1).
Catalog No. 50-233-1638