Learn More
Sino Biological Anti-Carbonic Anhydrase XIV/CA14 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50013RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CA14 extracellular domain (rM CA14; Catalog#50013-M08H; NP_035927.1; Met 1-Met 290). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3554