Learn More
Sino Biological Anti-Carbonic anhydrase II/CA2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50685T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Carbonic anhydrase II/CA2 (rM Carbonic anhydrase II/CA2; Catalog#50685-M08E; AAH55291.1; Met1-Lys260). Carbonic anhydrase II/CA2 specific IgG was purified by Mouse Carbonic anhydrase II/CA2 affinity chromatography.
Catalog No. 50-255-3154