Learn More
Sino Biological Anti-Carbonic Anhydrase III / CAIII Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107383T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse Carbonic Anhydrase III/CA3, and purified by antigen affinity chromatography.
Catalog No. 50-241-1262