Learn More
Sino Biological Anti-Carbonic Anhydrase IV/CA4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50350RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Carbonic Anhydrase IV / Car4 / CA4 (Catalog#50350-M08H; NP_031633.1; Met1-Ser277). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3300