Learn More
Sino Biological Recombinant Anti-Carbonic Anhydrase 8/CA8 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50767R05720
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase VIII / CA8 (rM Carbonic Anhydrase VIII / CA8; Catalog#50767-M08E; P28651; Met1-Gln291).
Catalog No. 50-255-3098