Learn More
Sino Biological Anti-Carbonic Anhydrase 8/CA8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50767RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Carbonic Anhydrase VIII / CA8 (Catalog#50767-M08E; P28651; Met1-Gln291). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3097