Learn More
Sino Biological Recombinant Mouse Carbonic Anhydrase IX/CA9 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50660M08HB20
Encompass_Preferred
A DNA sequence encoding the Mouse CA9 (NP_647466.2) (Met1-Asp390) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-3160