Learn More
ACROBiosystems Mouse Cadherin-17, His Tag (CA7-M52H5) is expressed from human 293 cells (HEK293). It contains AA Phe 26 - Met 786 (Accession # Q9R100-1).
Supplier:  ACROBiosystems CA7M52H5100UG
Encompass_Preferred
Cadherin-17, also known as liver-intestine (LI) Cadherin, belongs to the cadherin family of calcium-dependent cell adhesion molecules. In vivo studies showed CDH17 knockout resulted in apoptotic PC tumor death through activating caspase-3 activity. Taken together, CDH17 functions as an oncogenic molecule critical to PC growth by regulating tumor apoptosis signaling pathways and CDH17 could be targeted to develop an anti-PC therapeutic approach.
Catalog No. 50-223-1444