Learn More
ACROBiosystems Mouse Cadherin-17 25ug
Supplier:  ACROBiosystems CA7M82F325UG
Encompass_Preferred
Biotinylated Mouse Cadherin-17 Protein FcAvitag (CA7-M82F3) is expressed from human 293 cells (HEK293). It contains AA Phe 26 - Met 786 (Accession # Q9R100-1).
Catalog No. 50-253-5435