Learn More
Biomatik Corporation Mouse Cadherin, Heart (CDHH) ELISA Kit
Supplier:  Biomatik Corporation EKU02843
Encompass_Preferred
Mouse Cadherin, Heart (CDHH) ELISA Kit for detecting CDHH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.50-4000pg/ml. Sensitivity: 23.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WTR5. Gene ID: 12554. Target Synonyms: CDH13; P105; T-cad; Cadherin 13; T-Cadherin; H-Cadherin; Truncated cadherin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6132