Learn More
Sino Biological Anti-CADM3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50476RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CADM3 (rM CADM3; Catalog#50476-M08H; NP_444429.1; Met 1-His 328). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3260