Learn More
Sino Biological Anti-CADM3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50476T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CADM3/NECL1/IGSF4B (rM CADM3/NECL1/IGSF4B; Catalog#50476-M08H; NP_444429.1; Met1-His328). CADM3/NECL1/IGSF4B specific IgG was purified by Mouse CADM3/NECL1/IGSF4B affinity chromatography.
Catalog No. 50-255-3258