Learn More
Innovive Inc Mouse cage bottom for use with Innovive IVC or static mouse lids, prefilled with 1/8"""" corn cob bedding with single sheet paper enrichment added
Catalog No. NC1764786
Encompass_Preferred
Innocob (corn cob) is proven to encapsulate the urea crystals found in animal urine, controlling ammonia levels. Its sponge-like characteristics encapsulate ammonia and trap associated odors. PRE-BEDDED, CORN COB WITH 1-PLY INNORICHMENT
Catalog No. NC1764786 Supplier Innovive Inc Supplier No. MBTMC8E1
Innocob (corn cob) is proven to encapsulate the urea crystals found in animal urine, controlling ammonia levels. Its sponge-like characteristics encapsulate ammonia and trap associated odors. PRE-BEDDED, CORN COB WITH 1-PLY INNORICHMENT