Learn More
Biomatik Corporation Mouse Calbindin (CALB) ELISA Kit
Supplier:  Biomatik Corporation EKU10080
Encompass_Preferred
Mouse Calbindin (CALB) ELISA Kit for detecting CALB in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12658. Gene ID: 12307. Target Synonyms: CAB27; Calbindin 1,28kDa; Calbindin D28; Vitamin D-dependent calcium-binding protein, avian-type. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6097