Learn More
Biomatik Corporation Mouse Calcium Channel, Voltage Dependent, R-Type, Alpha 1E Subunit (CACNa1E) ELISA Kit
Supplier:  Biomatik Corporation EKU02866
Encompass_Preferred
Mouse Calcium Channel, Voltage Dependent, R-Type, Alpha 1E Subunit (CACNa1E) ELISA Kit for detecting CACNa1E in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61290. Target Synonyms: BII; CACNL1A6; CACH6; Brain calcium channel II; Calcium channel, L type, alpha-1 polypeptide, isoform 6; Voltage-gated calcium channel subunit alpha Cav2.3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5505