Learn More
Biomatik Corporation Mouse Calcium Sensing Receptor (CASR) ELISA Kit
Supplier:  Biomatik Corporation EKU02868
Encompass_Preferred
Mouse Calcium Sensing Receptor (CASR) ELISA Kit for detecting CASR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QY96. Gene ID: 12374. Target Synonyms: CAR; FHH; FIH; GPRC2A; HHC; HHC1; NSHPT; PCAR1; Parathyroid cell calcium-sensing receptor 1; Hypocalciuric Hypercalcemia 1; Severe Neonatal Hyperparathyroidism. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7239