Learn More
Biomatik Corporation Mouse Calcium/Calmodulin Dependent Protein Kinase II Alpha (CAMK2a) ELISA Kit
Supplier:  Biomatik Corporation EKU08359
Encompass_Preferred
Mouse Calcium/Calmodulin Dependent Protein Kinase II Alpha (CAMK2a) ELISA Kit for detecting CAMK2a in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11798. Gene ID: 12322. Target Synonyms: CAMKA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6930