Learn More
Biomatik Corporation Mouse Calpain 1 (CAPN1) ELISA Kit
Supplier:  Biomatik Corporation EKU02883
Encompass_Preferred
Mouse Calpain 1 (CAPN1) ELISA Kit for detecting CAPN1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35350. Gene ID: 12333. Target Synonyms: PIG30; CANPL1; MuCANP; MuCL; Calpain 1, Large Subunit; Calpain-1 Catalytic Subunit; Calcium-Dependent Protease 1; Calcium-activated neutral proteinase 1; Calpain mu-type; Micromolar-calpain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9084