Learn More
Biomatik Corporation Mouse Calpain 10 (CAPN10) ELISA Kit
Supplier:  Biomatik Corporation EKU02884
Encompass_Preferred
Mouse Calpain 10 (CAPN10) ELISA Kit for detecting CAPN10 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.103ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ESK3. Gene ID: 23830. Target Synonyms: Calcium-activated neutral proteinase 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9083