Learn More
Sino Biological Anti-CAMKI Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106746T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse CAMKI, and purified by antigen affinity chromatography.
Catalog No. 50-241-2275