Learn More
Sino Biological Recombinant Anti-CAMKIV Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50838R162100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CaM Kinase 4 / CaMKIV (Catalog#50838-M20B; P08414; Met1-Tyr469).
Catalog No. 50-240-9746