Learn More
Sino Biological Mouse CAMK4 PAb 100UL 50838-RP02-100
Supplier:  Sino Biological 50838RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CAMK4 / CaMKIV (rM CAMK4 / CaMKIV; Catalog#50838-M20B; P08414; Met1-Tyr469). CAMK4 / CaMKIV specific IgG was purified by Mouse CAMK4 / CaMKIV affinity chromatography.
Catalog No. 50-240-8913