Learn More
American Research Products Inc Mouse cAMP response element bi
Catalog No. 5000328541
Encompass
Mouse cAMP response element binding protein (CREB) ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-28541 Supplier American Research Products Inc Supplier No. CSBE15860M
Mouse cAMP response element binding protein (CREB) ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma cell culture supernates Method Sandwich